Merck
  • Proinflammatory effects of pancreatic elastase are mediated through TLR4 and NF-kappaB.
Proinflammatory effects of pancreatic elastase are mediated through TLR4 and NF-kappaB.
Biochemical and biophysical research communications (2004-09-08)
Antti Hietaranta, Harri Mustonen, Pauli Puolakkainen, Reijo Haapiainen, Esko Kemppainen
ABSTRACT
Pancreatic elastase has been implicated in the pathophysiology of severe acute pancreatitis, characterized by systemic inflammatory response, distant organ failure, and high mortality. Here we show that pancreatic elastase activates transcription factors NF-kappaB, AP-1, and NFAT in human myeloid cells (U-937 and THP-1) in culture. Pancreatic elastase also induces TNF-alpha secretion and increased expression of CD11b in THP-1 cells which can be inhibited by neutralizing anti-Toll-like receptor 4 (TLR4) antibodies. NF-kappaB blocking agents (MG-132, PGA1) prevented elastase-induced TNF-alpha secretion from THP-1 cells. Our results suggest that pancreatic elastase-induced proinflammatory effects are mediated by TLR4 and NF-kappaB in human myeloid cells.