Merck
Dnali1 is required for sperm motility and male fertility in mice.
Basic and clinical andrology (2023-11-23)
Yiling Zhou, Yaling Wang, Jingwen Chen, Bangguo Wu, Shuyan Tang, Feng Zhang, Chunyu Liu, Lingbo Wang
ABSTRACT
The sperm flagellum is an evolutionarily conserved specialized organelle responsible for sperm motility and male fertility. Deleterious mutations in genes involved in the sperm flagellum assembly can often cause sperm motility defects and male infertility. The murine Dnali1 gene encodes a protein that is known to interact with the cytoplasmic dynein heavy chain 1. A Dnali1-mutated mouse model was generated by inducing a nonsense mutation in the Dnali1 gene. The Dnali1-mutated male mice presented impaired sperm motility and were completely infertile. Although no obviously abnormal sperm morphology was observed in Dnali1-mutated male mice, the ultrastructural structure of sperm flagellum was disrupted, displaying as an asymmetrical distribution of the longitudinal columns (LCs). Notably, infertile Dnali1-mutated male mice were able to obtain offspring via ICSI. Our results uncover a role of DNALI1 in sperm motility and male fertility in mice, and demonstrate that ICSI overcomes Dnali1-associated male infertility, thus providing guidance for the diagnosis and genetic counseling of DNALI1-associated human infertility.
Sigma-Aldrich
Anti-DNAH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution