Merck
  • The BCL-2 family protein BCL-RAMBO interacts and cooperates with GRP75 to promote its apoptosis signaling pathway.
The BCL-2 family protein BCL-RAMBO interacts and cooperates with GRP75 to promote its apoptosis signaling pathway.
Scientific reports (2023-08-29)
Jinghong Xu, Takuya Hashino, Reiji Tanaka, Koichiro Kawaguchi, Hideki Yoshida, Takao Kataoka
ABSTRACT
The BCL-2 family protein BCL-RAMBO, also known as BCL2-like 13, anchors at the outer mitochondrial membrane and regulates apoptosis, mitochondrial fragmentation, and mitophagy. However, the mechanisms underlying the proapoptotic role of BCL-RAMBO remain unclear. In the present study, we demonstrated that BCL-RAMBO interacted with glucose-regulated protein 75 (GRP75), also known as heat shock protein family A member 9, and mortalin using co-immunoprecipitation and glutathione S-transferase-based pull-down assays. BCL-RAMBO interacted with GRP75 via its No BCL-2 homology domain. The interaction between BCL-RAMBO and GRP75 was confirmed by genetic interactions in Drosophila because a rough eye phenotype caused by the ectopic expression of BCL-RAMBO was partially suppressed by mutations in Hsc70-5, a mammalian GRP75 ortholog. In human embryonic kidney 293T cells, the co-expression of BCL-RAMBO and GRP75 facilitated an elevation in executioner caspase activity and poly (ADP-ribose) polymerase 1 (PARP-1) cleavage. In contrast, the knockdown of GRP75 suppressed elevated executioner caspase activity and PARP-1 cleavage in BCL-RAMBO-transfected cells. The mitochondrial release of cytochrome c induced by BCL-RAMBO was also attenuated by the knockdown of GRP75. These results indicate that GRP75 interacts with BCL-RAMBO and plays a crucial role in the BCL-RAMBO-dependent apoptosis signaling pathway.