Merck
  • Dynamics of TERT regulation via alternative splicing in stem cells and cancer cells.
Dynamics of TERT regulation via alternative splicing in stem cells and cancer cells.
PloS one (2023-08-02)
Jeongjin J Kim, Mohammed E Sayed, Alexander Ahn, Aaron L Slusher, Jeffrey Y Ying, Andrew T Ludlow
ABSTRACT
Part of the regulation of telomerase activity includes the alternative splicing (AS) of the catalytic subunit telomerase reverse transcriptase (TERT). Although a therapeutic window for telomerase/TERT inhibition exists between cancer cells and somatic cells, stem cells express TERT and rely on telomerase activity for physiological replacement of cells. Therefore, identifying differences in TERT regulation between stem cells and cancer cells is essential for developing telomerase inhibition-based cancer therapies that reduce damage to stem cells. In this study, we measured TERT splice variant expression and telomerase activity in induced pluripotent stem cells (iPSCs), neural progenitor cells (NPCs), and non-small cell lung cancer cells (NSCLC, Calu-6 cells). We observed that a NOVA1-PTBP1-PTBP2 axis regulates TERT alternative splicing (AS) in iPSCs and their differentiation into NPCs. We also found that splice-switching of TERT, which regulates telomerase activity, is induced by different cell densities in stem cells but not cancer cells. Lastly, we identified cell type-specific splicing factors that regulate TERT AS. Overall, our findings represent an important step forward in understanding the regulation of TERT AS in stem cells and cancer cells.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Anti-U2AF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody