Merck
  • Pericardial malignant mesothelioma diagnosed in a dog by immunocytochemistry of the pericardial fluid: a case report.
Pericardial malignant mesothelioma diagnosed in a dog by immunocytochemistry of the pericardial fluid: a case report.
BMC veterinary research (2023-07-22)
Eun Wha Choi
ABSTRACT
Pericardial effusions are one of the most common cardiac diseases in dogs. Common causes of haemorrhagic pericardial effusions include neoplasia, such as hemangiosarcoma, mesothelioma, chemodectoma, and ectopic thyroid tumours, and benign idiopathic pericardial effusion. Distinguishing among reactive mesothelial cells, malignant mesothelioma, and adenocarcinoma in body effusions is a diagnostic challenge. Therefore, the author aimed to discover whether the observed cells were reactive mesothelial, mesothelioma, or adenocarcinoma cells through immunocytochemistry using five markers (cytokeratin, vimentin, desmin, E-cadherin, and calretinin) in a canine patient. A 2.1 kg, spayed female, 10-year-old Yorkshire Terrier dog presented to a local hospital with dyspnoea and was evaluated for pericardial effusion. The presence of pericardial fluid was confirmed, and she was referred to our hospital for further evaluation. In cytological evaluation, cells shed individually or in clusters were observed, along with numerous non-degenerative neutrophils and macrophages. The cells showed binucleation, anisocytosis, anisokaryosis, abnormal nucleoli, abundant basophilic cytoplasm, high nuclear-cytoplasmic ratio, and coarse chromatin. Large atypical multinucleate cells were also observed. Erythrophagia was observed, indicating chronic haemorrhage. Immunocytochemistry using pericardial fluid was positive for cytokeratin, vimentin, desmin, E-cadherin, and calretinin. Therefore, malignant mesothelioma was diagnosed. Immunocytochemistry is a very useful diagnostic technique because it can determine whether several fluorescent markers are simultaneously expressed in the same cell. Further, E-cadherin and calretinin can be used for the differential diagnosis of reactive mesothelial cells, malignant mesothelioma, and adenocarcinoma in dogs.