Merck
  • Glypican-3 (GPC-3) Structural Analysis and Cargo in Serum Small Extracellular Vesicles of Hepatocellular Carcinoma Patients.
Glypican-3 (GPC-3) Structural Analysis and Cargo in Serum Small Extracellular Vesicles of Hepatocellular Carcinoma Patients.
International journal of molecular sciences (2023-07-14)
Montalbano Mauro, Perricone Ugo, Zachary Walton, Shirafkan Ali, Cristiana Rastellini, Luca Cicalese
ABSTRACT
Glypican-3 (GPC-3) is a heparin sulfate proteoglycan located extracellularly and anchored to the cell membrane of transformed hepatocytes. GPC-3 is not expressed in normal or cirrhotic liver tissue but is overexpressed in hepatocellular carcinoma (HCC). Because of this, GPC-3 is one of the most important emerging immunotargets for treatment and as an early detection marker of HCC. To determine if GPC-3 domains associated with serum small extracellular vesicles (sEVs) could be used as an HCC diagnostic marker, we predicted in silico GPC-3 structural properties and tested for the presence of its full-length form and/or cleaved domains in serum sEVs isolated from patients with HCC. Structural analysis revealed that the Furin cleavage site of GPC-3 is exposed and readily accessible, suggesting the facilitation of GPC-3 cleavage events. Upon isolation of sEVs from both hepatocytes, culture media and serum of patients with HCC were studied for GPC-3 content. This data suggests that Furin-dependent GPC-3 cleaved domains could be a powerful tool for detection of initial stages of HCC and serve as a predictor for disease prognosis.