Merck
  • A novel homozygous frameshift variant in DNAH8 causes multiple morphological abnormalities of the sperm flagella in a consanguineous Pakistani family.
A novel homozygous frameshift variant in DNAH8 causes multiple morphological abnormalities of the sperm flagella in a consanguineous Pakistani family.
Asian journal of andrology (2022-10-30)
Sobia Dil, Asad Khan, Ahsanullah Unar, Meng-Lei Yang, Imtiaz Ali, Aurang Zeb, Huan Zhang, Jian-Teng Zhou, Muhammad Zubair, Khalid Khan, Shun Bai, Qing-Hua Shi
ABSTRACT
Multiple morphological abnormalities of the sperm flagella (MMAF) is a severe form of asthenozoospermia categorized by immotile spermatozoa with abnormal flagella in ejaculate. Whole-exome sequencing (WES) is used to detect pathogenic variants in patients with MMAF. In this study, a novel homozygous frameshift variant (c.6158_6159insT) in dynein axonemal heavy chain 8 (DNAH8) from two infertile brothers with MMAF in a consanguineous Pakistani family was identified by WES. Reverse transcription-polymerase chain reaction (RT-PCR) confirmed DNAH8 mRNA decay in these patients with the DNAH8 mutation. Hematoxylin-eosin staining and transmission electron microscopy revealed highly divergent morphology and ultrastructure of sperm flagella in these patients. Furthermore, an immunofluorescence assay showed the absence of DNAH8 and a reduction in its associated protein DNAH17 in the patients' spermatozoa. Collectively, our study expands the phenotypic spectrum of patients with DNAH8-related MMAF worldwide.
Sigma-Aldrich
Anti-DNAH8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1