Merck
  • Huoxue Qianyang Qutan recipe attenuates Ang II-induced cardiomyocyte hypertrophy by regulating reactive oxygen species production.
Huoxue Qianyang Qutan recipe attenuates Ang II-induced cardiomyocyte hypertrophy by regulating reactive oxygen species production.
Experimental and therapeutic medicine (2021-11-02)
Mingtai Gui, Lei Yao, Bo Lu, Jing Wang, Xunjie Zhou, Jianhua Li, Zhenhua Dong, Deyu Fu
ABSTRACT
Continuous and irreversible cardiac hypertrophy can induce cardiac maladaptation and cardiac remodeling, resulting in increased risk of developing cardiovascular diseases. The present study was conducted to investigate the therapeutic effect of Huoxue Qianyang Qutan recipe (HQQR) on angiotensin II (Ang II)-induced cardiomyocyte hypertrophy. Primary cardiomyocytes were isolated from the cardiac tissue of neonatal rats, followed by flow cytometry detection to confirm the proportion of primary cardiomyocytes. Cell Counting Kit-8 assay and immunofluorescence detection were performed to examine the effect of Ang II and HQQR on cardiomyocyte hypertrophy. Reactive oxygen species (ROS) and a series of metabolic indicators were quantified to investigate the effect of HQQR on Ang II-induced cardiomyocyte hypertrophy. Mitochondrial electron transport chain complex activity and related coding gene expression were determined to explore the effect of HQQR on mitochondrial function. HQQR significantly inhibited Ang II-induced cardiomyocyte hypertrophy and restored Ang II-induced ROS accumulation, metabolic indicators, and membrane potential levels. HQQR also regulated the mitochondrial function related to the sirtuin 1 pathway in Ang II-induced cardiomyocytes by increasing the activity of the mitochondrial electron transport chain complex and affecting the expression of genes encoding mitochondrial electron transport chain complex subunits. HQQR could alleviate Ang II-induced cardiomyocyte hypertrophy by modulating oxidative stress, accumulating ROS and increasing mitochondrial electron transport chain activity.