- In vitro activities of polycationic peptides alone and in combination with clinically used antimicrobial agents against Rhodococcus equi.
In vitro activities of polycationic peptides alone and in combination with clinically used antimicrobial agents against Rhodococcus equi.
Antimicrobial agents and chemotherapy (1999-08-03)
A Giacometti, O Cirioni, F Ancarani, M S Del Prete, M Fortuna, G Scalise
PMID10428947
ABSTRACT
The in vitro activities of magainin II, nisin, and ranalexin alone and in combination with other antimicrobial agents against six clinical isolates of Rhodococcus equi were investigated by MIC and time-kill studies. All isolates were more susceptible to nisin. A positive interaction was observed when the peptides were combined with ampicillin, ceftriaxone, rifabutin, rifampin, azithromycin, clarithromycin, and vancomycin.