Merck
  • Inhibitory Activity of Quaternary Isoquinoline Alkaloids on Soluble Epoxide Hydrolase.
Inhibitory Activity of Quaternary Isoquinoline Alkaloids on Soluble Epoxide Hydrolase.
Current issues in molecular biology (2022-09-23)
Jang Hoon Kim, Chong Woon Cho, Mok Hur, Woo Tae Park, Youn-Ho Moon, Sung-Cheol Koo, Yun-Chan Hur, Jong Seong Kang, Ik Soo Lee
ABSTRACT
The quaternary isoquinoline alkaloids of palmatine (1), berberine (2), and jatrorrhizine (3) were evaluated in terms of their ability to inhibit soluble epoxide hydrolase (sEH). They had similar inhibitory activities, with IC50 values of 29.6 ± 0.5, 33.4 ± 0.8, and 27.3 ± 0.4 μM, respectively. Their respective Ki values of 26.9, 46.8, and 44.5 μM-determined by enzyme kinetics-indicated that they inhibited the catalytic reaction by binding noncompetitively with sEH. The application of computational chemistry to the in vitro results revealed the site of the receptor to which the ligand would likely bind. Accordingly, three alkaloids were identified as having a suitable basic skeleton for lead compound development of sEH inhibitors.
Sigma-Aldrich
Palmatine chloride, ≥98% (HPLC)