Merck
  • Synthesis of α-D-Ribose 1-Phosphate and 2-Deoxy-α-D-Ribose 1-Phosphate Via Enzymatic Phosphorolysis of 7-Methylguanosine and 7-Methyldeoxyguanosine.
Synthesis of α-D-Ribose 1-Phosphate and 2-Deoxy-α-D-Ribose 1-Phosphate Via Enzymatic Phosphorolysis of 7-Methylguanosine and 7-Methyldeoxyguanosine.
Current protocols (2022-01-21)
Irina V Varizhuk, Vladimir E Oslovsky, Pavel N Solyev, Mikhail S Drenichev, Sergey N Mikhailov
ABSTRACT
A simple and efficient method for the preparation of α-D-ribose 1-phosphate and 2-deoxy-α-D-ribose 1-phosphate, key intermediates in nucleoside metabolism and important starting compounds for the enzymatic synthesis of various modified nucleosides, has been proposed. It consists in near-irreversible enzymatic phosphorolysis of readily prepared hydroiodide salts of 7-methylguanosine and 7-methyl-2'-deoxyguanosine, respectively, in the presence of purine nucleoside phosphorylase. α-D-Ribose 1-phosphate and 2-deoxy-α-D-ribose 1-phosphate are obtained in near quantitative yields (by HPLC analysis) and 74%-94% yields after their isolation and purification. © 2022 Wiley Periodicals LLC. Basic Protocol 1: Preparation of α-D-ribose 1-phosphate barium salt (4a) Alternate Protocol 1: Preparation of 2-deoxy-α-D-ribose 1-phosphate barium salt (4b) Basic Protocol 2: Preparation of α-D-ribose 1-phosphate bis(cyclohexylammonium) salt (5a) Alternate Protocol 2: Preparation of 2-deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt (5b).