Merck
  • Differential effects of NT-3 on reinnervation of the fast extensor digitorum longus (EDL) and the slow soleus muscle of rat.
Differential effects of NT-3 on reinnervation of the fast extensor digitorum longus (EDL) and the slow soleus muscle of rat.
The European journal of neuroscience (2000-04-13)
M Simon, G Terenghi, C J Green, G R Coulton
ABSTRACT
Previous studies of gastrocnemius muscle reinnervation showed specific normalization of the proportion and diameter of fast type 2b muscle fibres following NT-3 delivery to the proximal stump of the cut sciatic nerve. Here, we investigate if normalization was related to greater improvement of muscle reinnervation of fast (extensor digitorum longus; EDL) than slow (soleus) motor units. NT-3-impregnated (NT-3 group) or plain fibronectin (FN group) mats were inserted into a sciatic nerve gap. Neuromuscular junctions (NMJs) labelled with TRITC-alpha-bungarotoxin were colabelled with calcitonin gene-related peptide (CGRP) or 4E2 antisera and imaged using confocal microscopy. CGRP and 4E2 were used as markers for newly reinnervated and structurally mature NMJs, respectively. At 40 days postsurgery, denervated NMJs in EDL and soleus muscles of both groups presented a 50% decrease of surface area due to decreased width. At day 80 in EDL, more NMJs were reinnervated by CGRP-immunoreactive terminals in the NT-3 (7.1%) than in the FN group (4.2%); there was no difference between groups for soleus. At 120 days, 4E2-immunoreactive NMJs were more numerous in EDL of the NT-3 (40.0%) than in the FN group (7.3%), unlike in soleus (NT-3, 1. 6%; FN, 1.8%), and presented a partial size recovery. These results indicate that NT-3 preferentially improves reinnervation of fast muscles over slow muscle, although the mechanism of this improvement is still unclear.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution