Merck
Analyzing Homologous Recombination at a Genome-Wide Level.
Methods in molecular biology (Clifton, N.J.) (2020-08-26)
Coline Arnould, Vincent Rocher, Gaëlle Legube
ABSTRACT
Among the types of damage, DNA double-strand breaks (DSBs) (provoked by various environmental stresses, but also during normal cell metabolic activity) are the most deleterious, as illustrated by the variety of human diseases associated with DSB repair defects. DSBs are repaired by two groups of pathways: homologous recombination (HR) and nonhomologous end joining. These pathways do not trigger the same mutational signatures, and multiple factors, such as cell cycle stage, the complexity of the lesion and also the genomic location, contribute to the choice between these repair pathways. To study the usage of the HR machinery at DSBs, we propose a genome-wide method based on the chromatin immunoprecipitation of the HR core component Rad51, followed by high-throughput sequencing.
Sigma-Aldrich
Anti-XRCC4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial