Merck
  • Whole-exome sequencing identified ARL2 as a novel candidate gene for MRCS (microcornea, rod-cone dystrophy, cataract, and posterior staphyloma) syndrome.
Whole-exome sequencing identified ARL2 as a novel candidate gene for MRCS (microcornea, rod-cone dystrophy, cataract, and posterior staphyloma) syndrome.
Clinical genetics (2019-04-05)
Xue-Bi Cai, Kun-Chao Wu, Xiao Zhang, Ji-Neng Lv, Guang-Hui Jin, Lue Xiang, Jie Chen, Xiu-Feng Huang, Deng Pan, Bin Lu, Fan Lu, Jia Qu, Zi-Bing Jin
ABSTRACT
Adenosine diphosphate (ADP)-ribosylation factor-like 2 (ARL2) protein participates in a broad range of cellular processes and acts as a mediator for mutant ARL2BP in cilium-associated retinitis pigmentosa and for mutant HRG4 in mitochondria-related photoreceptor degeneration. However, mutant ARL2 has not been linked to any human disease so far. Here, we identified a de novo variant in ARL2 (c.44G > T, p.R15L) in a Chinese pedigree with MRCS (microcornea, rod-cone dystrophy, cataract, and posterior staphyloma) syndrome through whole-exome sequencing and co-segregation analysis. Co-immunoprecipitation assay and immunoblotting confirmed that the mutant ARL2 protein showed a 62% lower binding affinity for HRG4 while a merely 18% lower binding affinity for ARL2BP. Immunofluorescence images of ARL2 and HRG4 co-localizing with cytochrome c in HeLa cells described their relationship with mitochondria. Further analyses of the mitochondrial respiratory chain and adenosine triphosphate production showed significant abnormalities under an ARL2-mutant condition. Finally, we generated transgenic mice to test the pathogenicity of this variant and observed retinal degeneration complicated with microcornea and cataract that were similar to those in our patients. In conclusion, we uncover ARL2 as a novel candidate gene for MRCS syndrome and suggest a mitochondria-related mechanism of the first ARL2 variant through site-directed mutagenesis studies.
Sigma-Aldrich
Anti-UNC119 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-ARL2 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution