Merck
  • SLAMF1 Is Dispensable for Vaccine-Induced T Cell Development but Required for Resistance to Fungal Infection.
SLAMF1 Is Dispensable for Vaccine-Induced T Cell Development but Required for Resistance to Fungal Infection.
Journal of immunology (Baltimore, Md. : 1950) (2022-02-27)
Elaine M Kohn, Lucas Dos Santos Dias, Hannah E Dobson, Xin He, Huafeng Wang, Bruce S Klein, Marcel Wüthrich
ABSTRACT
Homotypic signaling lymphocyte activation molecule (SLAM) receptor-ligand cell surface interactions between myeloid and lymphoid cells regulate innate and adaptive immune responses. In this article, we report that SLAMF1 is indispensable for host resistance to primary and vaccine-induced protection against fungal infection. Because vaccine immunity is dependent on cell-mediated immunity, we investigated the development of Ag-specific T cells. We studied the T cell-intrinsic and -extrinsic role of SLAMF1. We generated SLAMF1-/- TCR transgenic mice and analyzed the responses of adoptively transferred T cells. We also tracked endogenous Ag-specific T cells by using a tetramer. Intrinsic and extrinsic SLAMF1 signaling was dispensable for the development of antifungal Th1 and Th17 cells, which are requisite for the acquisition of vaccine-induced immunity. Despite intact T cell development, vaccinated SLAMF1-/- mice failed to control fungal infection. Failed accumulation of Ag-specific T cells in the lung on infection of vaccinated mice was due to uncontrolled early infection and inflammation, revealing a role for SLAMF1 in innate host immunity.