Merck
  • Reprogramming fibroblasts toward cardiomyocytes, neural stem cells and hepatocytes by cell activation and signaling-directed lineage conversion.
Reprogramming fibroblasts toward cardiomyocytes, neural stem cells and hepatocytes by cell activation and signaling-directed lineage conversion.
Nature protocols (2015-06-05)
Saiyong Zhu, Haixia Wang, Sheng Ding
ABSTRACT
Induction of tissue-specific cell types via a conventional transdifferentiation strategy typically uses overexpression of the corresponding lineage-specific transcription factors. Alternatively, somatic cells can be temporarily activated via a common set of reprogramming factors into a transition state, which can then be directed into various cell types via soluble lineage-specific signals, without establishing a pluripotent state. Here, we provide protocols for the generation of cardiomyocytes, neural stem cells and hepatocytes from fibroblasts with such a cell activation (CA) and signaling-directed (SD; CASD) strategy. In these protocols, beating cardiomyocytes can be induced from mouse fibroblasts in 2-5 weeks; expandable neural stem cells and definitive endoderm progenitors can be obtained from human fibroblasts as early as 2.5 weeks; and human definitive endoderm progenitors can be differentiated into functional hepatocytes in 2 weeks. Through further developments, the CASD strategy can serve as a unique avenue for generating diverse functional cell types for biomedical research and therapeutic applications.