Merck
  • Local application of platelet-activating factor induces cervical ripening accompanied by infiltration of polymorphonuclear leukocytes in rats.
Local application of platelet-activating factor induces cervical ripening accompanied by infiltration of polymorphonuclear leukocytes in rats.
American journal of obstetrics and gynecology (2002-10-22)
Holger Maul, Leili Shi, Stephen G Marx, Robert E Garfield, George R Saade
ABSTRACT
The purpose of this study was to determine the effect of locally applied platelet-activating factor on cervical ripening and leukocyte infiltration. Timed-pregnant rats were treated vaginally twice each day on days 14 and 15 of gestation with platelet-activating factor 5 x 10(-8) mol/L (low-dose), 5 x 10(-7) mol/L (high-dose), or solvent and were killed on day 16. Cervical ripening was assessed by cervical resistance-to-stretch and light-induced autofluorescence of cross-linked collagen. Longitudinal sections of the cervix were hematoxylin and eosin stained, and the number of polymorphonuclear leukocytes in eight high-power fields was determined. Cervical resistance-to-stretch and light-induced autofluorescence were decreased in rats that were treated with high-dose platelet-activating factor compared with controls (0.4375 +/- 0.0126 N/mm vs 0.5528 +/- 0.0294 N/mm, and 2.739 +/- 0.150 N/mm vs 3.475 +/- 0.236 N/mm; P <.05). High-dose platelet-activating factor led to infiltration with polymorphonuclear leukocytes compared with control (22.1 +/- 5.2 vs 2.1 +/- 1.3 cells/8 high-power fields, P <.05). Local platelet-activating factor application induces cervical ripening accompanied by stromal polymorphonuclear leukocyte infiltration. Platelet-activating factor may play a major role in parturition under physiologic and pathologic conditions.
Sigma-Aldrich
β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate, ≥98%