Merck
  • Effect of beta-muricholic acid on the prevention and dissolution of cholesterol gallstones in C57L/J mice.
Effect of beta-muricholic acid on the prevention and dissolution of cholesterol gallstones in C57L/J mice.
Journal of lipid research (2002-10-29)
David Q-H Wang, Susumu Tazuma
ABSTRACT
This study investigated whether beta-muricholic acid, a natural trihydroxy hydrophilic bile acid of rodents, acts as a biliary cholesterol-desaturating agent to prevent cholesterol gallstones and if it facilitates the dissolution of gallstones compared with ursodeoxycholic acid (UDCA). For gallstone prevention study, gallstone-susceptible male C57L mice were fed 8 weeks with a lithogenic diet (2% cholesterol and 0.5% cholic acid) with or without 0.5% UDCA or beta-muricholic acid. For gallstone dissolution study, additional groups of mice that have formed gallstones were fed chow with or without 0.5% beta-muricholic acid or UDCA for 8 weeks. One hundred percent of mice fed the lithogenic diet formed cholesterol gallstones. Addition of beta-muricholic acid and UDCA decreased gallstone prevalence to 20% and 50% through significantly reducing biliary secretion rate, saturation index, and intestinal absorption of cholesterol, as well as inducing phase boundary shift and an enlarged Region E that prevented the transition of cholesterol from its liquid crystalline phase to solid crystals and stones. Eight weeks of beta-muricholic acid and UDCA administration produced complete gallstone dissolution rates of 100% and 60% compared with the chow (10%). We conclude that beta-muricholic acid is more effective than UDCA in treating or preventing diet-induced or experimental cholesterol gallstones in mice.
Sigma-Aldrich
β-Muricholic Acid, ≥98% (HPLC)