Merck
  • Single-cell RNA sequencing reveals immunological rewiring at the maternal-fetal interface following asymptomatic/mild SARS-CoV-2 infection.
Single-cell RNA sequencing reveals immunological rewiring at the maternal-fetal interface following asymptomatic/mild SARS-CoV-2 infection.
Cell reports (2022-06-07)
Suhas Sureshchandra, Michael Z Zulu, Brianna M Doratt, Allen Jankeel, Delia Tifrea, Robert Edwards, Monica Rincon, Nicole E Marshall, Ilhem Messaoudi
ABSTRACT
While severe coronavirus 2019 (COVID-19) is associated with immune activation at the maternal-fetal interface, responses to asymptomatic/mild severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection during pregnancy remain unknown. Here, we assess immunological adaptations in blood and term decidua in response to asymptomatic/mild disease in pregnant women. We report attenuated antigen presentation and type I interferon (IFN) signaling pathways, loss of tissue-resident decidual macrophages, and upregulated cytokine/chemokine signaling in monocyte-derived decidual macrophages. Furthermore, we describe increased frequencies of activated tissue-resident T cells and decreased abundance of regulatory T cells with infection while frequencies of cytotoxic CD4/CD8 T cells are increased in the blood. In contrast to decidual macrophages, type I IFN signaling is higher in decidual T cells. Finally, infection leads to a narrowing of T cell receptor diversity in both blood and decidua. Collectively, these observations indicate that asymptomatic/mild COVID-19 during pregnancy results in remodeling of the immunological landscape of the maternal-fetal interface, with a potential for long-term adverse outcomes for the offspring.