Merck
  • SGSM2 inhibits thyroid cancer progression by activating RAP1 and enhancing competitive RAS inhibition.
SGSM2 inhibits thyroid cancer progression by activating RAP1 and enhancing competitive RAS inhibition.
Cell death & disease (2022-03-11)
Xi Su, Dong Chen, Lizhang Zhu, Hao Jia, Jiaxuan Cai, Peng Li, Bin Han, Donglai Wang, Hongtao Li, Jiaqian Fan, Mengwei Gu, Yaqi Zhou, Haixia Guan, Wei Wei
ABSTRACT
Thyroid cancer (TC) is one of the most common malignancies involving the head and neck, and its incidences are increasing every year. Small G protein signaling modulators 2 (SGSM2) belongs to a newly identified protein group that contributes to numerous cancer progression. However, its role in TC remains unknown. The aim of this study was to explore the functions and underlying molecular mechanism of SGSM2 in the progression of thyroid tumorigenesis. Here, we demonstrated that SGSM2 expression was markedly decreased in TC, and that lower SGSM2 expression was potentially related to worse patient prognosis. Meanwhile, the SGSM2 levels were not directly correlated with BRAF or RAS mutations in TC. Based on our functional analysis, ectopic SGSM2 expression strongly prevented cell proliferation, migration, invasion, and tumorigenic activity in TC cells that harbored wild type RAS. Mechanistically, we demonstrated that SGSM2 interacted with Small G protein Ras-associated protein 1(RAP1) and augmented its activity. Activated RAP1 then competitively suppressed RAS activation and thereby downregulated output of MAPK/ERK and PI3K/Akt networks, which are primary contributors of TC. In summary, the present study reports a tumor suppressive role of SGSM2 in TC. Moreover, we revealed the underlying molecular mechanism, thus providing a potential therapeutic target for TCs that harbor wild type RAS.