Merck
  • Investigation of an Outbreak of Dengue Virus Serotype 1 in a Rural Area of Kribi, South Cameroon: A Cross-Sectional Study.
Investigation of an Outbreak of Dengue Virus Serotype 1 in a Rural Area of Kribi, South Cameroon: A Cross-Sectional Study.
Intervirology (2019-05-03)
Fredy Brice Nemg Simo, Francine Berlange Sado Yousseu, Armel Evouna Mbarga, Jean Joel Bigna, Armel Melong, Anicet Ntoude, Basile Kamgang, Raphael Bouyne, Paul Moundipa Fewou, Maurice Demanou
ABSTRACT
On May 2017, a case of dengue serotype 1 was detected and confirmed through routine surveillance in a traveler returning from Kribi, a seaside town of Southern Cameroon. This study aimed at confirming the circulation of dengue virus (DENV) in Southern Cameroon. A cross sectional study was carried out in Londji near Kribi from June 21-25, 2017, by a joint team of Centre Pasteur of Cameroon and the Department of Diseases, Epidemics and Pandemics Control. Blood samples of consented participants were collected and tested for anti-D ENV IgM using an IgM antibody capture enzyme-linked immunosorbent assay (MAC-ELISA), and for the detection of Zika, dengue, or chikungunya viruses using Trioplex real-time reverse transcription-polymerase chain reaction (RT-PCR). DENV RNA-positive samples were serotyped using an end-point nested RT-PCR. Ninety-one participants were enrolled, 50.55% (46/91) of them males. The mean age of the population was 30.71 years (±18.89). In total, 14.28% (13/91) of the participants had DENV infection (3 anti-DENV IgM positive and 10 DENV serotype 1 RT-PCR positive). The detection of DENV serotype 1 in an autochthonous population during this survey is a confirmation that the seaside city of Kribi is a risk area for contracting dengue infection in Cameroon.