Merck
  • Regulation of bFGF-induced effects on rat aortic smooth muscle cells by β3-adrenergic receptors.
Regulation of bFGF-induced effects on rat aortic smooth muscle cells by β3-adrenergic receptors.
Current research in pharmacology and drug discovery (2022-03-19)
Yingzi Chang, Lei Alena Dagat, Aisha Yusuf, Yusuf Zahriya, Kotryna Staputyte, Emma Worley, Alex Holt, Natalie Canuteson, Vereena Messieha, Kasey Halila
ABSTRACT
Basic fibroblast growth factor (bFGF)-mediated vascular smooth muscle cell (VSMC) proliferation and migration play an important role in vascular injury-induced neointima formation and subsequent vascular restenosis, a major event that hinders the long-term success of angioplasty. The function of β3-adrenergic receptors (β3-ARs) in vascular injury-induced neointima formation has not yet been defined. Our current study explored the possible role of β3-ARs in vascular injury-induced neointima formation by testing its effects on bFGF-induced VSMC migration and proliferation. β3-AR expression in rat carotid arteries was examined at 14 days following a balloon catheter-induced injury. The effects of β3-AR activation on bFGF-induced rat aortic smooth muscle cell proliferation, migration, and signaling transduction (including extracellular-signal-regulated kinase/mitogen activated protein kinase, ERK/MAPK and Protein kinase B, AKT) were tested. We found that vascular injury induced upregulation of β3-ARs in neointima. Pretreatment of VSMCs with a selective β3-AR agonist, CL316,243 significantly potentiated bFGF-induced cell migration and proliferation, and ERK and AKT phosphorylation. Our results also revealed that suppressing phosphorylation of ERK and AKT blocked bFGF-induced cell migration and that inhibiting AKT phosphorylation reduced bFGF-mediated cell proliferation. Our results suggest that activation of β3-ARs potentiates bFGF-mediated effects on VSMCs by enhancing bFGF-mediated ERK and AKT phosphorylation and that β3-ARs may play a role in vascular injury-induced neointima formation.