Merck
  • Identification and characterization of a novel regulatory factor: IgA-inducing protein.
Identification and characterization of a novel regulatory factor: IgA-inducing protein.
Journal of immunology (Baltimore, Md. : 1950) (2003-07-23)
Amy S Austin, Karen M Haas, Sasha M Naugler, Anna A Bajer, David Garcia-Tapia, D Mark Estes
ABSTRACT
IgA is the predominant Ig isotype in mucosal secretions and thus plays a pivotal role in host defense. The mechanisms by which IgA expression is regulated may differ among species and involve multiple pathways. Various cytokines and costimulators have been identified which regulate expression of this isotype, including IL-10, IL-2, vasoactive intestinal peptide, and TGF-beta. We have tested a wide array of known factors, but only under very limited conditions do these factors mediate substantial IgA production in vitro from bovine B cells. In response to these findings, we generated a cDNA library in a mammalian expression vector from activated cells derived from bovine gut-associated lymphoid tissues (Peyer's patch and mesenteric lymph node cells) as a source of soluble factor(s) that may regulate IgA production. We have identified a novel factor, IgA-inducing protein, which stimulates relatively high levels of IgA production in vitro following CD40 stimulation in coculture with IL-2. Our data suggest that IgA-inducing protein regulates IgA by acting as a switch or differentiation factor and is expressed in a variety of lymphoid and nonlymphoid tissues.