Merck
  • Potential for reversing miR-634-mediated cytoprotective processes to improve efficacy of chemotherapy against oral squamous cell carcinoma.
Potential for reversing miR-634-mediated cytoprotective processes to improve efficacy of chemotherapy against oral squamous cell carcinoma.
Molecular therapy oncolytics (2022-05-17)
Phuong Xuan Tran, Jun Inoue, Hiroyuki Harada, Johji Inazawa
ABSTRACT
For advanced oral squamous cell carcinoma (OSCC), increasing sensitivity to chemotherapy is a major challenge in improving treatment outcomes, and targeting cytoprotective processes that lead to the chemotherapy resistance of cancer cells may be therapeutically promising. Tumor-suppressive microRNAs (miRNAs) can target multiple cancer-promoting genes concurrently and are thus expected to be useful seeds for cancer therapeutics. We revealed that miR-634-mediated targeting of multiple cytoprotective process-related genes, including cellular inhibitor of apoptosis protein 1 (cIAP1), can effectively increase cisplatin (CDDP)-induced cytotoxicity and overcome CDDP resistance in OSCC cells. The combination of topical treatment with miR-634 ointment and administration of CDDP was synergistically effective against OSCC tumor growth in a xenograft mouse model. Furthermore, the expression of miR-634 target genes is frequently upregulated in primary OSCC tumors. Our study suggests that reversing miR-634-mediated cytoprotective processes activated in cancer cells is a potentially useful strategy to improve CDDP efficacy against advanced OSCC.
Sigma-Aldrich
Anti-TFAM antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution