Merck
  • A Biomimetic Electrospun Membrane Supports the Differentiation and Maturation of Kidney Epithelium from Human Stem Cells.
A Biomimetic Electrospun Membrane Supports the Differentiation and Maturation of Kidney Epithelium from Human Stem Cells.
Bioengineering (Basel, Switzerland) (2022-05-28)
Xingrui Mou, Jessica Shah, Rohan Bhattacharya, Titilola D Kalejaiye, Bowen Sun, Po-Chun Hsu, Samira Musah
ABSTRACT
Podocytes derived from human induced pluripotent stem (hiPS) cells are enabling studies of kidney development and disease. However, many of these studies are carried out in traditional tissue culture plates that do not accurately recapitulate the molecular and mechanical features necessary for modeling tissue- and organ-level functionalities. Overcoming these limitations requires the design and application of tunable biomaterial scaffolds. Silk fibroin is an attractive biomaterial due to its biocompatibility and versatility, which include its ability to form hydrogels, sponge-like scaffolds, and electrospun fibers and membranes appropriate for tissue engineering and biomedical applications. In this study, we show that hiPS cells can be differentiated into post-mitotic kidney glomerular podocytes on electrospun silk fibroin membranes functionalized with laminin. The resulting podocytes remain viable and express high levels of podocyte-specific markers consistent with the mature cellular phenotype. The resulting podocytes were propagated for at least two weeks, enabling secondary cell-based applications and analyses. This study demonstrates for the first time that electrospun silk fibroin membrane can serve as a supportive biocompatible platform for human podocyte differentiation and propagation. We anticipate that the results of this study will pave the way for the use of electrospun membranes and other biomimetic scaffolds for kidney tissue engineering, including the development of co-culture systems and organs-on-chips microphysiological devices.