Merck
  • Luciferase assay system to monitor fibroblast growth factor signal disruption in human iPSCs.
Luciferase assay system to monitor fibroblast growth factor signal disruption in human iPSCs.
STAR protocols (2022-06-10)
Seiya Kanno, Kashu Mizota, Yusuke Okubo, Tatsuto Kageyama, Lei Yan, Junji Fukuda
ABSTRACT
We describe a protocol for a live-cell luciferase assay system for continuously monitoring fibroblast growth factor (FGF) signal disruption in human-induced pluripotent stem cells (iPSCs). Signal disrupting effects of chemicals are used as an indicator to evaluate toxicity. The assay is reliably predictive of the effects of limb malformation chemicals (AUC = 0.93). The current approach is limited to FGF signal disruption, and combinations with other types of signaling will be required to detect the effects of different toxicants. For complete details on the use and execution of this protocol, please refer to Kanno et al. (2022a).
Product Description
Roche
Agarose Gel DNA Extraction Kit, kit of for up to 100 reactions