- Light-Regulated NO Release as a Novel Strategy To Overcome Doxorubicin Multidrug Resistance.
Light-Regulated NO Release as a Novel Strategy To Overcome Doxorubicin Multidrug Resistance.
ACS medicinal chemistry letters (2017-03-25)
Konstantin Chegaev, Aurore Fraix, Elena Gazzano, Gamal Eldein F Abd-Ellatef, Marco Blangetti, Barbara Rolando, Sabrina Conoci, Chiara Riganti, Roberta Fruttero, Alberto Gasco, Salvatore Sortino
PMID28337331
ABSTRACT
Nitric oxide (NO) release from a suitable NO photodonor (NOP) can be fine-tuned by visible light stimuli at doses that are not toxic to cells but that inhibit several efflux pumps; these are mainly responsible for the multidrug resistance of the anticancer agent doxorubicin (DOX). The strategy may thus increase DOX toxicity against resistant cancer cells. Moreover, a novel molecular hybrid covalently joining DOX and NOP showed similar increased toxicity toward resistant cancer cells and, in addition, lower cardiotoxicity than DOX. This opens new and underexplored approaches to overcoming the main therapeutic drawbacks of this chemotherapeutic based on light-controlled release of NO.