Merck
  • Resistin and IL-15 as Predictors of Invasive Mechanical Ventilation in COVID-19 Pneumonia Irrespective of the Presence of Obesity and Metabolic Syndrome.
Resistin and IL-15 as Predictors of Invasive Mechanical Ventilation in COVID-19 Pneumonia Irrespective of the Presence of Obesity and Metabolic Syndrome.
Journal of personalized medicine (2022-03-26)
Carles Perpiñan, Laia Bertran, Ximena Terra, Carmen Aguilar, Jessica Binetti, Miguel Lopez-Dupla, Anna Rull, Laia Reverté, Elena Yeregui, Frederic Gómez-Bertomeu, Joaquim Peraire, Teresa Auguet, On Behalf Of Covid-Study Group
ABSTRACT
The cytokine signature present in COVID-19 could provide information on the pathogenic mechanisms of the disease and could identify possible prognostic biomarkers and possible therapeutic targets. In this longitudinal work, we studied the clinical and biochemical parameters and circulating cytokine levels of 146 patients at the time of admission for COVID-19 and 4-6 weeks later. The main objective of this study was to determine whether basal cytokines could be early prognostic biomarkers of COVID-19, and also to analyze the impact of comorbidities, such as obesity or metabolic syndrome (MS), in the cytokine profile. The levels of most inflammatory cytokines were elevated on admission in relation to the level that was reached 4-6 weeks later, except for IL-1β, which was lower on admission; these levels were irrespective of the presence of obesity or MS since the cytokine storm masks these inflammatory processes. Among the cytokines analyzed, those that correlated with a worse prognosis of COVID-19 were resistin, IL-6, IL-8, IL-15, MCP-1 and TNF-α. Specifically, resistin and IL-15 are the best early predictors of requiring invasive ventilation. Therefore, resistin and IL-15 should be included in the personalized treatment decision algorithm of patients with COVID-19.