Merck
  • Development and validation of a method for the analysis of five diamide insecticides in edible mushrooms using modified QuEChERS and HPLC-MS/MS.
Development and validation of a method for the analysis of five diamide insecticides in edible mushrooms using modified QuEChERS and HPLC-MS/MS.
Food chemistry (2020-07-14)
Fajun Tian, Chengkui Qiao, Jing Luo, Linlin Guo, Tao Pang, Rongli Pang, Jun Li, Caixia Wang, Ruiping Wang, Hanzhong Xie
ABSTRACT
In this study, a new method for simultaneous determination of cyantraniliprole, chlorantraniliprole, tetrachlorantraniliprole, cyclaniliprole and flubendiamide in edible mushrooms by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) combined with a modified QuEChERS procedure. The samples were extracted using acetonitrile and then cleaned up by primary secondary amine (PSA) and octadecylsilane (C18). The determination of these insecticides was achieved in less than 5 min using an electrospray ionization source in positive mode (ESI+) for cyantraniliprole and chlorantraniliprole, while negative mode (ESI-) for tetrachlorantraniliprole, cyclaniliprole and flubendiamide. The linearities of the calibrations for all target compounds were acceptable (R2 ≥ 0.9922). The limits of detection and quantification were 0.05-2 μg kg-1 and 5 μg kg-1, respectively. Acceptable recoveries (73.5-110.2%) were acquired for these insecticides with RSDs less than 12.7%. The results demonstrated that the proposed method was effective and convenient for the determination of these insecticides in edible mushrooms.
Product Description
Supelco
Cyantraniliprole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland