Merck
  • The Nap family proteins, CG5017/Hanabi and Nap1, are essential for Drosophila spermiogenesis.
The Nap family proteins, CG5017/Hanabi and Nap1, are essential for Drosophila spermiogenesis.
FEBS letters (2013-03-05)
Shuhei Kimura
ABSTRACT
Spermiogenesis is a dynamic process leading to alterations in cell morphology. In spermiogenesis, the roles of the histone chaperones are largely unknown. Here, I report the unexpected roles of two Nap family proteins, CG5017/Hanabi and nucleosome assembly protein 1 (Nap1) in Drosophila. Hanabi is mainly localized in the cytoplasm, and the hanabi mutant shows fully scattered nuclei and abnormality of nuclear shaping in spermatid elongation. In contrast, Nap1 is localized at the apical tip of the sperm head, and the nap1 mutant exhibits disruption of the nuclear bundle in the later stage. These findings imply that Nap family proteins might individually sustain cytoskeleton-based morphogenesis, rather than histone biogenesis.
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine and potassium bicarbonate., liquid, sterile-filtered, suitable for insect cell culture