Merck
  • Closing the serological gap: promising novel biomarkers for the early diagnosis of rheumatoid arthritis.
Closing the serological gap: promising novel biomarkers for the early diagnosis of rheumatoid arthritis.
Autoimmunity reviews (2012-06-06)
Leendert A Trouw, Michael Mahler
ABSTRACT
Rheumatoid arthritis (RA) is a chronic autoimmune disease characterized by inflammation and damage of the joints affecting about 0.5% of the general population. Early treatment in RA is important as it can prevent disease progression and irreversible damage of the joints. Despite the high diagnostic value of anti-citrullinated protein antibodies (ACPA) and rheumatoid factor (RF), there is a strong demand for novel serological biomarkers to further improve the diagnosis of this abundant disease. During the last decades, several autoantigens have been described in RA including Ra33 (hnRNP A2), fibrinogen, fibronectin, alpha-enolase, type II collagen, immunoglobulin binding protein (BiP), annexins and viral citrullinated peptide (VCP) derived from Epstein Barr Virus-encoded protein (EBNA-2). More recent discoveries include antibodies to carbamylated antigens (anti-CarP), to peptidyl arginine deiminase type 4 (PAD4), to BRAF (v raf murine sarcoma viral oncogene homologue B1) and to 14 autoantigens identified by phage display technology. This review provides a current overview of novel biomarkers for RA and discusses their future potential to improve the diagnosis of the disease.