- Infestation and hydraulic consequences of induced carbon starvation.
Infestation and hydraulic consequences of induced carbon starvation.
Drought impacts on forests, including widespread die-off, are likely to increase with future climate change, although the physiological responses of trees to lethal drought are poorly understood. In particular, in situ examinations of carbon starvation and its interactions with and effects on infestation and hydraulic vulnerability are largely lacking. In this study, we conducted a controlled, in situ, repeated defoliation experiment to induce carbon stress in isolated trembling aspen (Populus tremuloides) ramets. We monitored leaf morphology, leaves per branch, and multitissue carbohydrate concentrations during canopy defoliation. We examined the subsequent effects of defoliation and defoliation-induced carbon stress on vulnerability to insect/fungus infestation and hydraulic vulnerability the following year. Defoliated ramets flushed multiple canopies, which coincided with moderate drawdown of nonstructural carbohydrate reserves. Infestation frequency greatly increased and hydraulic conductivity decreased 1 year after defoliation. Despite incomplete carbohydrate drawdown from defoliation and relatively rapid carbohydrate recovery, suggesting considerable carbohydrate reserves in aspen, defoliation-induced carbon stress held significant consequences for vulnerability to mortality agents and hydraulic performance. Our results indicate that multiyear consequences of drought via feedbacks are likely important for understanding forests' responses to drought and climate change over the coming decades.