- Targeted delivery of atorvastatin via asialoglycoprotein receptor (ASGPR).
Targeted delivery of atorvastatin via asialoglycoprotein receptor (ASGPR).
Bioorganic & medicinal chemistry (2019-04-22)
Youxi Zhang, Xinfu Zhang, Chunxi Zeng, Bin Li, Chengxiang Zhang, Wenqing Li, Xucheng Hou, Yizhou Dong
PMID31005367
ABSTRACT
Targeted drug delivery platforms can increase the concentration of drugs in specific cell populations, reduce adverse effects, and hence improve the therapeutic effect of drugs. Herein, we designed two conjugates by installing the targeting ligand GalNAc (N-acetylgalactosamine) onto atorvastatin (AT). Compared to the parent drug, these two conjugates, termed G2-AT and G2-K-AT, showed increased hepatic cellular uptake. Moreover, both conjugates were able to release atorvastatin, and consequently showed dramatic inhibition of β-hydroxy-β-methylglutaryl-CoA (HMG-CoA) reductase and increased LDL receptors on cell surface.