Merck
  • Myelin oligodendrocyte-specific protein is expressed in Müller cells of myelinated vertebrate retinas.
Myelin oligodendrocyte-specific protein is expressed in Müller cells of myelinated vertebrate retinas.
Journal of neuroscience research (2011-02-22)
Adela Quesada, Yolanda Aguilera, Rosalía Caparrós, Francisco A Prada, Concepción Santano, Rosario López-López, Carmen Prada
ABSTRACT
The retina of nonmammalian vertebrates has a loose myelin that enwraps the large axons of the ganglion cells in all areas, whereas that of mammals lacks myelin, with some exceptions, such as the rabbit retina, which shows compact myelin restricted to the myelinated streak. Electron microscopy studies in chicken retina showed processes of Müller cells (MCs) and oligodendrocytes enwrapping ganglion cell axons. How each of these cells contributes to chicken retina myelination and whether the MC of other myelinated retinas is involved in myelination remain unknown. By immunohistochemistry, with a monoclonal antibody against myelin oligodendrocyte-specific protein (MOSP), we show that MOSP is intensely expressed in the MC and the optic-fiber layer (OFL) in myelinated but not in unmyelinated retinas. By immunocytochemistry with isolated MCs from the chick and rabbit retinas, we show that MOSP is concentrated in the innermost domain of the vitread processes. By immunoblotting, we show that protein extracts from myelinated retinas, but not those from unmyelinated retinas, presented a single band labelled with anti-MOSP of molecular weight similar to that of brain MOSP. In addition, we show that the MC of the embryonic chicken retina starts to express MOSP just before myelination starts. Our results agree with those of electron microscopy studies showing myelin in chick retina formed by MC processes and with those of immunohistochemistry studies in rabbit and human retinas showing expression of other myelin molecules in the MC. Altogether, our results suggest that the MC in myelinated retinas might contribute MOSP to myelin.