Merck
  • Parity-induced changes to mammary epithelial cells control NKT cell expansion and mammary oncogenesis.
Parity-induced changes to mammary epithelial cells control NKT cell expansion and mammary oncogenesis.
Cell reports (2021-12-09)
Amritha Varshini Hanasoge Somasundara, Matthew A Moss, Mary J Feigman, Chen Chen, Samantha L Cyrill, Michael F Ciccone, Marygrace C Trousdell, Macy Vollbrecht, Siran Li, Jude Kendall, Semir Beyaz, John E Wilkinson, Camila O Dos Santos
ABSTRACT
Pregnancy reprograms mammary epithelial cells (MECs) to control their responses to pregnancy hormone re-exposure and carcinoma progression. However, the influence of pregnancy on the mammary microenvironment is less clear. Here, we used single-cell RNA sequencing to profile the composition of epithelial and non-epithelial cells in mammary tissue from nulliparous and parous female mice. Our analysis indicates an expansion of γδ natural killer T-like immune cells (NKTs) following pregnancy and upregulation of immune signaling molecules in post-pregnancy MECs. We show that expansion of NKTs following pregnancy is due to elevated expression of the antigen-presenting molecule CD1d on MECs. Loss of CD1d expression on post-pregnancy MECs, or overall lack of activated NKTs, results in mammary oncogenesis. Collectively, our findings illustrate how pregnancy-induced changes modulate the communication between MECs and the immune microenvironment and establish a causal link between pregnancy, the immune microenvironment, and mammary oncogenesis.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride