Merck
  • ACE2-like carboxypeptidase B38-CAP protects from SARS-CoV-2-induced lung injury.
ACE2-like carboxypeptidase B38-CAP protects from SARS-CoV-2-induced lung injury.
Nature communications (2021-11-25)
Tomokazu Yamaguchi, Midori Hoshizaki, Takafumi Minato, Satoru Nirasawa, Masamitsu N Asaka, Mayumi Niiyama, Masaki Imai, Akihiko Uda, Jasper Fuk-Woo Chan, Saori Takahashi, Jianbo An, Akari Saku, Ryota Nukiwa, Daichi Utsumi, Maki Kiso, Atsuhiro Yasuhara, Vincent Kwok-Man Poon, Chris Chung-Sing Chan, Yuji Fujino, Satoru Motoyama, Satoshi Nagata, Josef M Penninger, Haruhiko Kamada, Kwok-Yung Yuen, Wataru Kamitani, Ken Maeda, Yoshihiro Kawaoka, Yasuhiro Yasutomi, Yumiko Imai, Keiji Kuba
ABSTRACT
Angiotensin-converting enzyme 2 (ACE2) is a receptor for cell entry of SARS-CoV-2, and recombinant soluble ACE2 protein inhibits SARS-CoV-2 infection as a decoy. ACE2 is a carboxypeptidase that degrades angiotensin II, thereby improving the pathologies of cardiovascular disease or acute lung injury. Here we show that B38-CAP, an ACE2-like enzyme, is protective against SARS-CoV-2-induced lung injury. Endogenous ACE2 expression is downregulated in the lungs of SARS-CoV-2-infected hamsters, leading to elevation of angiotensin II levels. Recombinant Spike also downregulates ACE2 expression and worsens the symptoms of acid-induced lung injury. B38-CAP does not neutralize cell entry of SARS-CoV-2. However, B38-CAP treatment improves the pathologies of Spike-augmented acid-induced lung injury. In SARS-CoV-2-infected hamsters or human ACE2 transgenic mice, B38-CAP significantly improves lung edema and pathologies of lung injury. These results provide the first in vivo evidence that increasing ACE2-like enzymatic activity is a potential therapeutic strategy to alleviate lung pathologies in COVID-19 patients.