Merck
  • CRTC2 activation in the suprachiasmatic nucleus, but not paraventricular nucleus, varies in a diurnal fashion and increases with nighttime light exposure.
CRTC2 activation in the suprachiasmatic nucleus, but not paraventricular nucleus, varies in a diurnal fashion and increases with nighttime light exposure.
American journal of physiology. Cell physiology (2014-08-01)
Julie A Highland, Michael J Weiser, Laura R Hinds, Robert L Spencer
ABSTRACT
Entrainment of the intrinsic suprachiasmatic nucleus (SCN) molecular clock to the light-dark cycle depends on photic-driven intracellular signal transduction responses of SCN neurons that converge on cAMP response element-binding protein (CREB)-mediated regulation of gene transcription. Characterization of the CREB coactivator proteins CREB-regulated transcriptional coactivators (CRTCs) has revealed a greater degree of differential activity-dependent modulation of CREB transactivational function than previously appreciated. In confirmation of recent reports, we found an enrichment of crtc2 mRNA and prominent CRTC2 protein expression within the SCN of adult male rats. With use of a hypothalamic organotypic culture preparation for initial CRTC2-reactive antibody characterization, we found that CRTC2 immunoreactivity in hypothalamic neurons shifted from a predominantly cytoplasmic profile under basal culture conditions to a primarily nuclear localization (CRTC2 activation) 30 min after adenylate cyclase stimulation. In adult rat SCN, we found a diurnal variation in CRTC2 activation (peak at zeitgeber time of 4 h and trough at zeitgeber time of 16-20 h) but no variation in the total number of CRTC2-immunoreactive cells. There was no diurnal variation of CRTC2 activation in the hypothalamic paraventricular nucleus, another site of enriched CRTC2 expression. Exposure of rats to light (50 lux) for 30 min during the second half of their dark (nighttime) phase produced CRTC2 activation. We observed in the SCN a parallel change in the expression of a CREB-regulated gene (FOS). In contrast, nighttime light exposure had no effect on CRTC2 activation or FOS expression in the paraventricular nucleus, nor did it affect corticosterone hormone levels. These results suggest that CRTC2 participates in CREB-dependent photic entrainment of SCN function.