Merck
  • Sarcocystis neurona, seroprevalence of antibodies in equines and research of oocystis in opossum in Ilhéus - Itabuna microregion, Bahia, Brazil.
Sarcocystis neurona, seroprevalence of antibodies in equines and research of oocystis in opossum in Ilhéus - Itabuna microregion, Bahia, Brazil.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria (2021-06-24)
Samantha Gusmão Pellizzoni, Sônia Carmen Lopo Costa, Raissa Barros Gracie Mery, Jonata Melo Barbieri, Alexandre Dias Munhoz, Aísla Nascimento da Silva, Anaiá da Paixão Sevá, Martín Roberto Del Valle Alvarez, George Rêgo Albuquerque
ABSTRACT
The aims of this study were to determine the seroprevalence of Sarcocystis neurona antibodies in equines in the Ilhéus-Itabuna microregion (BA), and identify possible factors associated with infection. The presence of sporocysts/oocysts of Sarcocystis spp. was also verified in Didelphis spp. A total of 669 serum samples were collected from equines in 56 properties located in 12 municipalities in the region. Indirect fluorescent antibody test was performed with slides containing merozoites of the S. neurona, using a cut-off titer of 1:80. Occurrence of 7.92% of anti-S. neurona antibodies was observed in the sampled equines. The purposes trade and work were significantly associated with the presence of antibodies (p<0.05), and being used for the purpose of work (21.6%) was considered a risk factor, while being used for the purpose of trade (3.6%) was a protective factor. A total of 25 Didelphis spp. was captured for research on sporocysts/oocysts in stool samples and intestinal scrapings, being all negative. Didelphis spp. were all negative for the presence of Sarcocystis spp. and this circumstance does not change the fact that seroprevalence of S. neurona has been observed in horses raised in the southern Bahia.