Merck
  • Cancer-associated fibroblasts educate normal fibroblasts to facilitate cancer cell spreading and T-cell suppression.
Cancer-associated fibroblasts educate normal fibroblasts to facilitate cancer cell spreading and T-cell suppression.
Molecular oncology (2021-08-12)
Go Itoh, Kurara Takagane, Yuma Fukushi, Sei Kuriyama, Michinobu Umakoshi, Akiteru Goto, Kazuyoshi Yanagihara, Masakazu Yashiro, Masamitsu Tanaka
ABSTRACT
In some tumors, a small number of cancer cells are scattered in a large fibrotic stroma. Here, we demonstrate a novel mechanism for expansion of pro-tumor fibroblasts via cancer-associated fibroblast (CAF)-mediated education of normal fibroblasts (NFs). When NFs were incubated with conditioned medium from CAFs, the resulting CAF-educated fibroblasts (CEFs) generated reactive oxygen species, which induced NF-κB-mediated expression of inflammatory cytokines and the extracellular matrix protein asporin (ASPN), while expression of a common CAF marker gene, α-SMA, was not increased. ASPN further increased CEF expression of downstream molecules, including indoleamine 2,3-dioxygenase 1 (IDO-1), kynureninase (KYNU), and pregnancy-associated plasma protein-A (PAPP-A). These CEFs induce cytocidal effects against CD8+ T cells and IGF-I activation in cancer cells. CEFs were generated without cancer cells by the direct mixture of NFs and CAFs in mouse xenografts, and once CEFs were generated, they sequentially educated NFs, leading to continuous generation of CEFs. In diffuse-type gastric cancers, ASPNhigh /IDO-1high /KYNUhigh /α-SMA- CEFs were located at the distal invading front. These CEFs expanded in the fibrotic stroma and caused dissemination of cancer cells. ASPN may therefore be a key molecule in facilitating tumor spreading and T-cell suppression.
Sigma-Aldrich
Anti-LIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KYNU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution