Merck
  • Protein interactome of the Cancerous Inhibitor of protein phosphatase 2A (CIP2A) in Th17 cells.
Protein interactome of the Cancerous Inhibitor of protein phosphatase 2A (CIP2A) in Th17 cells.
Current research in immunology (2021-04-06)
Mohd Moin Khan, Tommi Välikangas, Meraj Hasan Khan, Robert Moulder, Ubaid Ullah, Santosh Dilip Bhosale, Elina Komsi, Umar Butt, Xi Qiao, Jukka Westermarck, Laura L Elo, Riitta Lahesmaa
ABSTRACT
Cancerous inhibitor of protein phosphatase 2A (CIP2A) is involved in immune response, cancer progression, and Alzheimer's disease. However, an understanding of the mechanistic basis of its function in this wide spectrum of physiological and pathological processes is limited due to its poorly characterized interaction networks. Here we present the first systematic characterization of the CIP2A interactome by affinity-purification mass spectrometry combined with validation by selected reaction monitoring targeted mass spectrometry (SRM-MS) analysis in T helper (Th) 17 (Th17) cells. In addition to the known regulatory subunits of protein phosphatase 2A (PP2A), the catalytic subunits of protein PP2A were found to be interacting with CIP2A. Furthermore, the regulatory (PPP1R18, and PPP1R12A) and catalytic (PPP1CA) subunits of phosphatase PP1 were identified among the top novel CIP2A interactors. Evaluation of the ontologies associated with the proteins in this interactome revealed that they were linked with RNA metabolic processing and splicing, protein traffic, cytoskeleton regulation and ubiquitin-mediated protein degradation processes. Taken together, this network of protein-protein interactions will be important for understanding and further exploring the biological processes and mechanisms regulated by CIP2A both in physiological and pathological conditions.
Sigma-Aldrich
MS RT Calibration Mix, Proteomics Retention Time Standard for LC-MS