Merck
  • Discovery of ERD-308 as a Highly Potent Proteolysis Targeting Chimera (PROTAC) Degrader of Estrogen Receptor (ER).
Discovery of ERD-308 as a Highly Potent Proteolysis Targeting Chimera (PROTAC) Degrader of Estrogen Receptor (ER).
Journal of medicinal chemistry (2019-04-17)
Jiantao Hu, Biao Hu, Mingliang Wang, Fuming Xu, Bukeyan Miao, Chao-Yie Yang, Mi Wang, Zhaomin Liu, Daniel F Hayes, Krishnapriya Chinnaswamy, James Delproposto, Jeanne Stuckey, Shaomeng Wang
ABSTRACT
The estrogen receptor (ER) is a validated target for the treatment of estrogen receptor-positive (ER+) breast cancer. Here, we describe the design, synthesis, and extensive structure-activity relationship (SAR) studies of small-molecule ERα degraders based on the proteolysis targeting chimeras (PROTAC) concept. Our efforts have resulted in the discovery of highly potent and effective PROTAC ER degraders, as exemplified by ERD-308 (32). ERD-308 achieves DC50 (concentration causing 50% of protein degradation) values of 0.17 and 0.43 nM in MCF-7 and T47D ER+ breast cancer cell lines, respectively, and induces >95% of ER degradation at concentrations as low as 5 nM in both cell lines. Significantly, ERD-308 induces more complete ER degradation than fulvestrant, the only approved selective ER degrader (SERD), and is more effective in inhibition of cell proliferation than fulvestrant in MCF-7 cells. Further optimization of ERD-308 may lead to a new therapy for advanced ER+ breast cancer.
Sigma-Aldrich
(S,R,S)-AHPC-Me, 95%