Merck
  • Semaphorin 4A antibody alleviates arsenic-induced hepatotoxicity in mice via inhibition of AKT2/NF-κB inflammatory signaling.
Semaphorin 4A antibody alleviates arsenic-induced hepatotoxicity in mice via inhibition of AKT2/NF-κB inflammatory signaling.
Toxicology and applied pharmacology (2020-12-09)
Yuan Yang, Qinling Wang, Wenjuan Wang, Shaofeng Wei, Qibing Zeng, Aihua Zhang
ABSTRACT
Semaphorin (Sema) 3A and Sema 4A are immunomodulatory molecules with a common receptor, neuropilin-1 (NRP-1), on the immune cells. Sema 3A binds to NRP-1 and inhibits T cell activation and inflammation, while Sema 4A binds to NRP-1 and promotes T cell activation and inflammation. These molecules are associated closely with the regulation of protein kinase B (AKT)/nuclear factor-kappaB (NF-κB) signaling, which are poorly understood in arsenic toxicity. The present study explored the role of Sema 3A or Sema 4A in arsenic-induced hepatotoxicity in mice. Arsenic exposure induced hepatic injury and resulted in the activations of p-AKT2, NF-κB p65, and NOD-like receptor family pyrin domain-containing 3 (NLRP3) inflammasome, downregulation of Sema 3A, and upregulation of Sema 4A or NRP-1. Interestingly, intervention with anti-Sema 4A antibody showed the mitigation of arsenic-induced hepatotoxicity, accompanied by the downregulation of Sema 4A, rebound of Sema 3A, and upregulation of NRP-1. And, the inflammatory signaling p-AKT2 or NF-κB p65, and NLRP3 inflammasome showed a downregulation compared with arsenic treatment group. In contrast, anti-Sema 3A antibody intervention did not show the significant effect in the histopathological features compared with arsenic treatment group. In conclusion, the anti-Sema 4A antibody antagonizes arsenic-induced hepatotoxicity in mice and may be involved in the inhibitions of AKT2/NF-κB and NLRP3 inflammatory signaling mediated synergistically by Sema 4A or Sema 3A and their receptor NRP-1.