Merck
Inhibitors of histone demethylases.
Bioorganic & medicinal chemistry (2011-05-21)
Brian Lohse, Jesper L Kristensen, Line H Kristensen, Karl Agger, Kristian Helin, Michael Gajhede, Rasmus P Clausen
ABSTRACT
Methylated lysines are important epigenetic marks. The enzymes involved in demethylation have recently been discovered and found to be involved in cancer development and progression. Despite the relative recent discovery of these enzymes a number of inhibitors have already appeared. Most of the inhibitors are either previously reported inhibitors of related enzymes or compounds derived from these. Development in terms of selectivity and potency is still pertinent. Several reports on the development of functional assays have been published.
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Baicalein, 98%