- Topical bromfenac reduces multiple inflammatory cytokines in the aqueous humour of pseudophakic patients.
Topical bromfenac reduces multiple inflammatory cytokines in the aqueous humour of pseudophakic patients.
Intraocular surgery is associated with increased ocular inflammation. If maintained for a prolonged period after surgery, this inflammation can cause various complications, including subconjunctival fibrosis and bleb scarring. This clinical trial was a prospective, randomised, single-blind, interventional study comparing the efficacy and safety of 0.1% bromfenac sodium ophthalmic solution and 0.02% fluorometholone ophthalmic suspension in the inhibition of multiple inflammatory cytokines in the aqueous humour of 26 patients with pseudophakic eyes who had undergone phacoemulsification and intraocular lens implantation. The patients were randomly assigned to one of the trial drugs, and aqueous humour samples were collected before and after drug administration. Platelet-derived growth factor-AA levels significantly decreased in both drug groups, but they were significantly higher in the fluorometholone group than in the bromfenac group (P = 0.034). Bromfenac also significantly decreased vascular endothelial growth factor level (P = 0.0077), as well as monocyte chemoattractant protein-1 level (P = 0.013), which was elevated for a prolonged period after phacoemulsification. These data suggest that bromfenac is useful to alleviate prolonged microenvironmental alterations in the aqueous humour of pseudophakic eyes.