Merck
  • Honey and Chamomile Activate Keratinocyte Antioxidative Responses via the KEAP1/NRF2 System.
Honey and Chamomile Activate Keratinocyte Antioxidative Responses via the KEAP1/NRF2 System.
Clinical, cosmetic and investigational dermatology (2020-09-29)
Tatsuya Ogawa, Yosuke Ishitsuka, Yoshiyuki Nakamura, Naoko Okiyama, Rei Watanabe, Yasuhiro Fujisawa, Manabu Fujimoto
ABSTRACT
The stratum corneum protects against the entry of pathogens, allergens, and irritants while preventing dehydration. The Kelch-like erythroid cell-derived protein with cap-n-collar homology-associated protein 1 (KEAP1)/NF-E2-related factor 2 (NRF2) system maintains skin barrier homeostasis. Aggregated evidence suggests that NRF2-mediated antioxidative response is hardwired into the stratified squamous epithelia. Honey and chamomile have long been regarded as natural antioxidants. Nonetheless, it is still unclear whether they activate the KEAP1/NRF2 system in the epidermis and could promote epidermal barrier recovery. To address the abovementioned issue, we explored the antioxidative property of honey/chamomile extract by using non-cell-based KEAP1-inhibition assay and cultured human epidermal keratinocytes. Herein we report that the extract inhibited KEAP1-NRF2 interaction and induced keratinocyte production of antioxidant small proline-rich protein. Our results may offer an opportunity to develop cosmetic products that boost NRF2-mediated antioxidative/antiaging, epidermis-intrinsic bio-responses.
Sigma-Aldrich
tert-Butylhydroquinone, 97%