Merck
  • Hydroxyurea generates nitric oxide in human erythroid cells: mechanisms for gamma-globin gene activation.
Hydroxyurea generates nitric oxide in human erythroid cells: mechanisms for gamma-globin gene activation.
Experimental biology and medicine (Maywood, N.J.) (2009-08-07)
Tzu-Fang Lou, Manisha Singh, Ashley Mackie, Wei Li, Betty S Pace
ABSTRACT
Hydroxyurea (HU) induces fetal hemoglobin synthesis through activation of cyclic guanine monophosphate (cGMP) signaling. Studies in sickle cell patients demonstrated increased circulating nitric oxide (NO) levels after oral HU treatment. However, the direct measurement of NO in erythroid cells and its role in fetal hemoglobin induction have not been defined. Therefore, we quantified the level of nitrate and nitrite (NOx) generated by HU in human erythroid progenitors in the presence of three nitric oxide synthase inhibitors (NOS), including N(G)-monomethyl-L-arginine (L-NMMA). In addition, cGMP levels were measured in the presence or absence of the pathway inhibitor 1H-(1,2,4)ox-adiazolo(4,3-a)quinoxalin-1-one, which blocks soluble guanylyl cyclase formation. HU treatment increased NOx levels and gamma-globin transcription in K562 and primary erythroid cells, which was augmented when HU was combined with L-NMMA. Pretreatment with the cGMP pathway inhibitor reversed gamma-gene activation by HU. These data demonstrate the direct stimulation of cellular NO and cGMP signaling in erythroid progenitors by HU as a possible mechanism for gamma-globin gene activation.