Merck
  • Apoptosis of Ascogregarina taiwanensis (Apicomplexa: Lecudinidae), which failed to migrate within its natural host.
Apoptosis of Ascogregarina taiwanensis (Apicomplexa: Lecudinidae), which failed to migrate within its natural host.
The Journal of experimental biology (2012-09-22)
Wei-June Chen, Ching-Gi Huang, Mei-Huei Fan-Chiang, Yu-Han Liu, Yi-Feng Lee
ABSTRACT
Sexual reproduction of Ascogregarina taiwanensis (Apicomplexa: Lecudinidae), a parasite specific to the mosquito Aedes albopictus, in Malpighian tubules is initiated by the entry of the trophotozoites developed in the midgut shortly after pupation (usually <5 h). However, only a low proportion of trophozoites are able to migrate; others end up dying. In this study, we demonstrated that those trophozoites that failed to migrate eventually died of apoptosis. Morphological changes such as shrinkage, chromatin aggregations and formation of blunt ridges on the surface were seen in moribund trophozoites. In addition, DNA fragmentation of trophozoites isolated from the midgut of pupae was demonstrated by the presence of DNA ladders, Annexin V staining and TUNEL assays. Detection of caspase-like activity suggests that apoptosis of those trophozoites may have occurred through a mechanism of an intrinsic or mitochondrial-mediated pathway. Although apoptosis has been observed in various protozoan species, it is not clear how apoptosis in single-celled organisms might result from evolution by natural selection. However, we speculate that apoptosis may regulate the parasite load of A. taiwanensis within its natural mosquito host, leading to an optimized state of the survival rate for both parasite and host.