Merck
  • Beta-3 adrenergic stimulation of L-type Ca(2+) channels in rat portal vein myocytes.
Beta-3 adrenergic stimulation of L-type Ca(2+) channels in rat portal vein myocytes.
British journal of pharmacology (2000-04-01)
P Viard, N Macrez, F Coussin, J L Morel, J Mironneau
ABSTRACT
1. The effects of beta(3)-adrenergic stimulation were studied on the L-type Ca(2+) channel in single myocytes from rat portal vein using the whole-cell mode of the patch-clamp technique. 2. Reverse transcription-polymerase chain reaction showed that beta(1)-, beta(2)- and beta(3)-adrenoceptor subtypes were expressed in rat portal vein myocytes. Application of both propranolol (a non-selective beta(1)- and beta(2)-adrenoceptor antagonist) and SR59230A (a beta(3)-adrenoceptor antagonist) were needed to inhibit the isoprenaline-induced increase in L-type Ca(2+) channel current. 3. L-type Ca(2+) channels were stimulated by CGP12177A (a beta(3)-adrenoceptor agonist with potent beta(1)- and beta(2)-adrenoceptor antagonist property) in a manner similar to that of isoprenaline. The CGP12177A-induced stimulation of Ca(2+) channel current was blocked by SR59230A, cyclic AMP-dependent protein kinase inhibitors, H-89 and Rp 8-Br-cyclic AMPs, but was unaffected by protein kinase C inhibitors, GF109203X and 19-31 peptide. This stimulation was mimicked by forskolin and 8-Br-cyclic AMP. In the presence of okadaic acid (a phosphatase inhibitor), the beta(3)-adrenoceptor-induced stimulation was maintained after withdrawal of the agonist. 4. The beta(3)-adrenoceptor stimulation of L-type Ca(2+) channels was blocked by a pretreatment with cholera toxin and by the intracellular application of an anti-Galpha(s) antibody. This stimulation was unaffected by intracellular infusion of an anti-Gbeta(com) antibody and a betaARK(1) peptide. 5. These results show that activation of beta(3)-adrenoceptors stimulates L-type Ca(2+) channels in vascular myocytes through a Galpha(s)-induced stimulation of the cyclic AMP/protein kinase A pathway and the subsequent phosphorylation of the channels.