Merck
  • Prognostic value of association of OCT4 with LEF1 expression in esophageal squamous cell carcinoma and their impact on epithelial-mesenchymal transition, invasion, and migration.
Prognostic value of association of OCT4 with LEF1 expression in esophageal squamous cell carcinoma and their impact on epithelial-mesenchymal transition, invasion, and migration.
Cancer medicine (2018-07-06)
Yue Zhao, Chunguang Li, Lei Huang, Shuai Niu, Qijue Lu, Dejun Gong, Shengdong Huang, Yang Yuan, Hezhong Chen
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is a malignant disease with poor prognosis. Because of early metastasis prior to diagnosis and therapeutic resistance, ESCC has become one of the leading causes of cancer-related death. Here, we investigated the clinicopathological significance of the association of octamer-binding transcription factor 4 (OCT4) with lymphoid enhancer-binding factor 1 (LEF1) expression and the potential molecular mechanism in the epithelial-mesenchymal transition (EMT), invasion, and migration of ESCC. The expression of OCT4 and LEF1 was detected via immunohistochemistry analysis. High levels of LEF1 expression were observed in 95 ESCC specimens and were obviously associated with aberrant clinicopathological features and poor patient prognosis. Our previous study showed that OCT4 expression level is elevated in ESCC, and statistical analysis showed that the elevated expression of OCT4 and LEF1 in ESCC was significantly associated with histologic grade, lymph node metastasis, TNM stage, and poor patient prognosis. The specific inhibition of OCT4 expression via a lentivirus encoding OCT4-shRNA (LV-shOCT4) in Eca109 cells led to decreased levels of OCT4 and LEF1 in vitro. Additionally, we applied a rescue strategy by infecting LV-shOCT4 Eca109 cells with a LEF1 overexpression plasmid (p-LEF1) and detected changes in EMT, migration, and invasion. Unsurprisingly, the p-LEF1 group exhibited greater EMT, invasion, and migration than did the LV-shOCT4 and negative control groups. This study demonstrates for the first time the relationship between OCT4 and LEF1 expression. The combination of high expression of OCT4 and LEF1 was associated with clinicopathological features of atypical patients, and this combination might be an ideal prognostic factor in ESCC. OCT4 positively regulated LEF1 expression, and LEF1 mediated the effects of OCT4 in cancer cell EMT, invasion, and migration. The data presented here suggest that the inhibition of OCT4-LEF1 signaling may be a new therapeutic target for the treatment of ESCC.