Merck
  • Gi3 mediates somatostatin-induced activation of an inwardly rectifying K+ current in human growth hormone-secreting adenoma cells.
Gi3 mediates somatostatin-induced activation of an inwardly rectifying K+ current in human growth hormone-secreting adenoma cells.
Endocrinology (1997-06-01)
K Takano, J Yasufuku-Takano, A Teramoto, T Fujita
ABSTRACT
SRIF activates an inwardly rectifying K+ current in human GH-secreting adenoma cells. Activation of this K+ current induces hyperpolarization of the membrane and abolishment of action potential firing. This mechanism is an essential mechanism for SRIF-induced decrease in intracellular Ca2+ concentration and inhibition of GH secretion. The activation of the inwardly rectifying K+ current is mediated by a pertussis toxin-sensitive G protein. In this article, the expression of the pertussis toxin-sensitive G protein alpha-subunits in the human GH-secreting adenoma cells were analyzed by RT-PCR, and the G protein transducing the SRIF-induced activation of this inwardly rectifying K+ current was investigated. RT-PCR of the messenger RNA from two human GH-secreting adenomas revealed that all G alpha(i1), G alpha(i2), G alpha(i3), and G alpha(o) were expressed in these adenomas. Primary cultured cells from these two adenoma cells were investigated under the voltage clamp of the whole-cell mode. Specific antibodies against the carboxyl terminus of G protein alpha-subunits were microinjected into the cells. Microinjection of antibody against the carboxyl terminal sequence of G alpha(i3) attenuated the SRIF-induced activation of the inwardly rectifying K+ current, whereas antibody against the common carboxyl terminal sequence of G alpha(i1) and G alpha(i2) did not. These data indicate that the G protein transducing the SRIF-induced activation of the inwardly rectifying K+ current is Gi3.